missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Difluorobenzylamine, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC311530050

 View more versions of this product

Catalog No. AC311530050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

3, 4-Difluorobenzylamine
79°C
Glass bottle
1.21
5g
72235-53-1
100.0
C7H7F2N
MFCD00010145
PHLZUDXEBCQHKM-UHFFFAOYSA-N
(3,4-difluorophenyl)methanamine
123572
Liquid
1.2100g/mL
Authentic
1.4920 to 1.4940
Colorless to Yellow
97%
96.0
94% min. (GC)
F2C6H3CH2NH2
1-3,4-difluorophenyl methanamine, 3, 4-difluorobenzylamine, 3,4-difluoro-benzylamine, 3,4-difluorobenzyl amine, 3,4-difluorobenzylamine, 3,4-difluorophenyl methanamine, 3,4-difluorophenyl methylamine, acmc-209omb, benzenemethanamine, 3,4-difluoro, pubchem1918
C1=CC(=C(C=C1CN)F)F
143.14
143.14
95%

Chemical Identifiers

72235-53-1
143.14
PHLZUDXEBCQHKM-UHFFFAOYSA-N
123572
C1=CC(=C(C=C1CN)F)F
C7H7F2N
MFCD00010145
1-3,4-difluorophenyl methanamine, 3, 4-difluorobenzylamine, 3,4-difluoro-benzylamine, 3,4-difluorobenzyl amine, 3,4-difluorobenzylamine, 3,4-difluorophenyl methanamine, 3,4-difluorophenyl methylamine, acmc-209omb, benzenemethanamine, 3,4-difluoro, pubchem1918
(3,4-difluorophenyl)methanamine
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 276-503-6

Recommended Storage : May darken during storage

SDS Documents