Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Difluorobenzyl bromide, 98%

Chemical Identifiers

Chemical Name or Material 3,4-Difluorobenzyl bromide
CAS 85118-01-0
Molecular Formula C7H5BrF2
Formula Weight 207.02
MDL Number MFCD00009889
UN Number UN1760
Beilstein 742578
Synonym alpha-Bromo-3,4-difluorotoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1996509

  SDS
alfa aesar™
A1996509
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 207.02
Chemical Name or Material 3,4-Difluorobenzyl bromide
Boiling Point 59° to 61°C (3.5mmHg)
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Danger
Molecular Formula C7H5BrF2
UN Number UN1760
Assay 98%
Flash Point 79°C (174°F)
CAS 85118-01-0
Health Hazard 2 GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Beilstein 742578
Synonym alpha-Bromo-3,4-difluorotoluene
Quantity 10g
MDL Number MFCD00009889
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.