missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Difluorobenzonitrile, 97%, Maybridge

Chemical Identifiers

CAS 64248-62-0
Molecular Formula C7H3F2N
Molecular Weight (g/mol) 139.105
MDL Number MFCD00011666
InChI Key BTBFCBQZFMQBNT-UHFFFAOYSA-N
Synonym 3,4 difluorobenzonitrile
PubChem CID 587203
IUPAC Name 3,4-difluorobenzonitrile
SMILES C1=CC(=C(C=C1C#N)F)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
TL00656EA SDS View Documents Fisher Scientific Company
TL00656EA
10g Amber Glass Bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

64248-62-0
139.105
BTBFCBQZFMQBNT-UHFFFAOYSA-N
587203
C1=CC(=C(C=C1C#N)F)F
C7H3F2N
MFCD00011666
3,4 difluorobenzonitrile
3,4-difluorobenzonitrile

Specifications

Amber Glass Bottle
64248-62-0
97%
F2C6H3CN
3,4 difluorobenzonitrile
C1=CC(=C(C=C1C#N)F)F
139.105
139.1
3, 4-Difluorobenzonitrile
10g
97.0
C7H3F2N
MFCD00011666
BTBFCBQZFMQBNT-UHFFFAOYSA-N
3,4-difluorobenzonitrile
587203
97%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.