missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Difluorobenzenesulfonamide, 97%

Manufacturer:  Alfa Aesar™ H5594703

 View more versions of this product

Catalog No. AAH5594703



Specifications

Specifications

3,4-Difluorobenzenesulfonamide
1g
C6H5F2NO2S
3,4-difluoro benzene sulfonamide
C1=CC(=C(C=C1S(=O)(=O)N)F)F
193.168
193.17
89°C to 93°C
108966-71-8
MFCD00728800
VFVVRYNJTGHAIE-UHFFFAOYSA-N
3,4-difluorobenzenesulfonamide
822865
97%

Chemical Identifiers

108966-71-8
193.168
VFVVRYNJTGHAIE-UHFFFAOYSA-N
822865
C1=CC(=C(C=C1S(=O)(=O)N)F)F
C6H5F2NO2S
MFCD00728800
3,4-difluoro benzene sulfonamide
3,4-difluorobenzenesulfonamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents