missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Difluorobenzeneboronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H6202203

 View more versions of this product

Catalog No. AAH6202203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3,4-Difluorobenzeneboronic acid pinacol ester
754226-39-6
1,3,2-dioxaborolane, 2-3,4-difluorophenyl-4,4,5,5-tetramethyl
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)F
240.057
240.05
96%
1g
C12H15BF2O2
VJTCPZZOSKBSCP-UHFFFAOYSA-N
2-(3,4-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
53429043
Solid

Chemical Identifiers

754226-39-6
240.057
1,3,2-dioxaborolane, 2-3,4-difluorophenyl-4,4,5,5-tetramethyl
2-(3,4-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
C12H15BF2O2
VJTCPZZOSKBSCP-UHFFFAOYSA-N
53429043
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)F
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents