Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Difluorobenzeneboronic acid pinacol ester, 96%

Chemical Identifiers

Chemical Name or Material 3,4-Difluorobenzeneboronic acid pinacol ester
Formula Weight 240.05
Synonym 3,4- Difluorophenylboronic acid pinacol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6202203

 
alfa aesar™
H6202203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62022MD

 
alfa aesar™
H62022MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 240.05
Chemical Name or Material 3,4-Difluorobenzeneboronic acid pinacol ester
Quantity 1g
Assay 96%
Physical Form Solid
Synonym 3,4- Difluorophenylboronic acid pinacol ester