missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Difluorobenzamidoxime, 97%

Manufacturer:  Alfa Aesar™ H5463603

 View more versions of this product

Catalog No. AAH5463603



Specifications

Specifications

3,4-Difluorobenzamidoxime
500024-74-8
MFCD06660194
3,4-difluoro-n'-hydroxybenzene-1-carboximidamide
C1=CC(=C(C=C1C(=NO)N)F)F
172.135
172.13
1g
C7H6F2N2O
19347401
OGCLGUHODKKLSH-UHFFFAOYSA-N
3,4-difluoro-N'-hydroxybenzenecarboximidamide
17750753
97%

Chemical Identifiers

500024-74-8
172.135
OGCLGUHODKKLSH-UHFFFAOYSA-N
17750753
C1=CC(=C(C=C1C(=NO)N)F)F
C7H6F2N2O
MFCD06660194
3,4-difluoro-n'-hydroxybenzene-1-carboximidamide
3,4-difluoro-N'-hydroxybenzenecarboximidamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents