Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Difluorobenzaldehyde, 98%

$74.65 - $237.05

Chemical Identifiers

Chemical Name or Material 3,4-Difluorobenzaldehyde
CAS 34036-07-2
Molecular Formula C7H4F2O
Formula Weight 142.11
MDL Number MFCD00010328
Beilstein 2241231
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1402806

 
alfa aesar™
A1402806
5g Each for $74.65

AAA1402814

 
alfa aesar™
A1402814
25g Each for $237.05
Description & Specifications

Specifications

Formula Weight 142.11
Chemical Name or Material 3,4-Difluorobenzaldehyde
Assay 98%
Molecular Formula C7H4F2O
MDL Number MFCD00010328
CAS 34036-07-2
Beilstein 2241231
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H227-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P301+P310-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 69°C (156°F)
Boiling Point 180°C
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P210-P261-P301+P310-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H227-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.