missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Difluorobenzaldehyde, 96%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 271300050

 View more versions of this product

Catalog No. AC271300050



Description

Description

Specifications

Specifications

3, 4-Difluorobenzaldehyde
65°C
Glass bottle
1.29
5g
34036-07-2
100.0
C7H4F2O
MFCD00010328
JPHKMYXKNKLNDF-UHFFFAOYSA-N
3,4-difluorobenzaldehyde
588088
Liquid
1.2900g/mL
Authentic
1.4990 to 1.5010
Colorless to Yellow
96%
95.0
95% min. (GC)
F2C6H3CHO
3, 4-difluorobenzaldehyde, 3,4 difluorobenzaldehyde, 3,4-diflourobenzaldehyde, 3,4-difluoro benzaldehyde, 3,4-difluoro-benzaldehye, 3,4-difluro-benzaldehyde, 3.4-difluorobenzaldehyde, acmc-1ajc9, benzaldehyde, 3,4-difluoro, pubchem1413
C1=CC(=C(C=C1C=O)F)F
142.1
142.1
96%

Chemical Identifiers

34036-07-2
142.1
JPHKMYXKNKLNDF-UHFFFAOYSA-N
588088
C1=CC(=C(C=C1C=O)F)F
C7H4F2O
MFCD00010328
3, 4-difluorobenzaldehyde, 3,4 difluorobenzaldehyde, 3,4-diflourobenzaldehyde, 3,4-difluoro benzaldehyde, 3,4-difluoro-benzaldehye, 3,4-difluro-benzaldehyde, 3.4-difluorobenzaldehyde, acmc-1ajc9, benzaldehyde, 3,4-difluoro, pubchem1413
3,4-difluorobenzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

TSCA : TSCA

SDS Documents