Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Difluoroaniline, 98+%

$33.86 - $33.86

Chemical Identifiers

Chemical Name or Material 3,4-Difluoroaniline
CAS 3863-11-4
Molecular Formula C6H5F2N
Formula Weight 129.11
MDL Number MFCD00007761
UN Number UN2941
Beilstein 971235
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1399906

 
alfa aesar™
A1399906*
5g Each for $33.86

AAA1399914

 
alfa aesar™
A1399914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1399922

 
alfa aesar™
A1399922
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 129.11
Chemical Name or Material 3,4-Difluoroaniline
Assay ≥98%
Molecular Formula C6H5F2N
MDL Number MFCD00007761
CAS 3863-11-4
UN Number UN2941
Beilstein 971235
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H311-H373-H227-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: FLUOROANILINES
RTECS Number CX9871900
Flash Point 85°C (185°F)
Odor Characteristic
Boiling Point 196° to 198°C
Melting Point 20° to 22°C
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

GHS H Statement
H311-H373-H227-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.