missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Difluoro-N-(4-methylbenzyl)benzenesulfonamide, 97%

Chemical Identifiers

CAS 321705-77-5
Molecular Formula C14H13F2NO2S
Molecular Weight (g/mol) 297.32
MDL Number MFCD02556071
InChI Key LUPVVMZLWZQPGV-UHFFFAOYSA-N
Synonym 3,4-difluoro-n-4-methylphenyl methyl benzenesulfonamide, 3,4-difluoro-n-4-methylbenzyl benzenesulfonamide, 3,4-difluorophenyl sulfonyl 4-methylphenyl methyl amine, 3,4-difluoro-n-4-methylphenyl methyl benzene-1-sulfonamide
PubChem CID 6467917
IUPAC Name 3,4-difluoro-N-[(4-methylphenyl)methyl]benzenesulfonamide
SMILES CC1=CC=C(C=C1)CNS(=O)(=O)C2=CC(=C(C=C2)F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH61926MD View Documents Alfa Aesar™
H61926MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6192603 View Documents Alfa Aesar™
H6192603
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

321705-77-5
297.32
LUPVVMZLWZQPGV-UHFFFAOYSA-N
6467917
CC1=CC=C(C=C1)CNS(=O)(=O)C2=CC(=C(C=C2)F)F
C14H13F2NO2S
MFCD02556071
3,4-difluoro-n-4-methylphenyl methyl benzenesulfonamide, 3,4-difluoro-n-4-methylbenzyl benzenesulfonamide, 3,4-difluorophenyl sulfonyl 4-methylphenyl methyl amine, 3,4-difluoro-n-4-methylphenyl methyl benzene-1-sulfonamide
3,4-difluoro-N-[(4-methylphenyl)methyl]benzenesulfonamide

Specifications

250mg
C14H13F2NO2S
3,4-difluoro-n-4-methylphenyl methyl benzenesulfonamide, 3,4-difluoro-n-4-methylbenzyl benzenesulfonamide, 3,4-difluorophenyl sulfonyl 4-methylphenyl methyl amine, 3,4-difluoro-n-4-methylphenyl methyl benzene-1-sulfonamide
CC1=CC=C(C=C1)CNS(=O)(=O)C2=CC(=C(C=C2)F)F
297.32
297.32
3,4-Difluoro-N-(4-methylbenzyl)benzenesulfonamide
321705-77-5
MFCD02556071
LUPVVMZLWZQPGV-UHFFFAOYSA-N
3,4-difluoro-N-[(4-methylphenyl)methyl]benzenesulfonamide
6467917
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning