missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Difluoro-N-(4-fluorobenzyl)benzenesulfonamide, 97%

Chemical Identifiers

CAS 321705-45-7
Molecular Formula C13H10F3NO2S
Molecular Weight (g/mol) 301.283
MDL Number MFCD02556043
InChI Key AFXHLFLQECWKRV-UHFFFAOYSA-N
PubChem CID 7790813
IUPAC Name 3,4-difluoro-N-[(4-fluorophenyl)methyl]benzenesulfonamide
SMILES C1=CC(=CC=C1CNS(=O)(=O)C2=CC(=C(C=C2)F)F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH61745MD View Documents Alfa Aesar™
H61745MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6174503 View Documents Alfa Aesar™
H6174503
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

321705-45-7
301.283
AFXHLFLQECWKRV-UHFFFAOYSA-N
3,4-difluoro-N-[(4-fluorophenyl)methyl]benzenesulfonamide
C13H10F3NO2S
MFCD02556043
7790813
C1=CC(=CC=C1CNS(=O)(=O)C2=CC(=C(C=C2)F)F)F

Specifications

250mg
C13H10F3NO2S
AFXHLFLQECWKRV-UHFFFAOYSA-N
3,4-difluoro-N-[(4-fluorophenyl)methyl]benzenesulfonamide
7790813
97%
321705-45-7
MFCD02556043
C1=CC(=CC=C1CNS(=O)(=O)C2=CC(=C(C=C2)F)F)F
301.283
301.28
3,4-Difluoro-N-(4-fluorobenzyl)benzenesulfonamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning