Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Difluoro-L-phenylalanine, 98%

$159.72 - $399.30

Chemical Identifiers

Chemical Name or Material 3,4-Difluoro-L-phenylalanine
CAS 31105-90-5
Molecular Formula C9H9F2NO2
Formula Weight 201.18
MDL Number MFCD01075125
Synonym H-Phe(3,4-DiF)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2753903

  SDS
alfa aesar™
H2753903
1g Each for $399.30

AAH27539MD

  SDS
alfa aesar™
H27539MD
250mg Each for $159.72
Description & Specifications

Specifications

Formula Weight 201.18
Chemical Name or Material 3,4-Difluoro-L-phenylalanine
Quantity 1g
Molecular Formula C9H9F2NO2
CAS 31105-90-5
Synonym H-Phe(3,4-DiF)-OH
MDL Number MFCD01075125
Assay 98%