Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Difluoro-DL-phenylalanine, 97%

$50.47 - $50.47

Chemical Identifiers

Chemical Name or Material 3,4-Difluoro-DL-phenylalanine
CAS 32133-36-1
Molecular Formula C9H9F2NO2
Formula Weight 201.18
MDL Number MFCD00061307
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH32276MD

  SDS
alfa aesar™
H32276MD
250mg Each for $50.47
Description & Specifications

Specifications

Formula Weight 201.18
Molecular Formula C9H9F2NO2
Quantity 250mg
Assay 97%
MDL Number MFCD00061307
Melting Point 232° to 234°C
Chemical Name or Material 3,4-Difluoro-DL-phenylalanine
CAS 32133-36-1