Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Difluoro-5-methylbenzaldehyde, 97%

$82.35 - $275.53

Chemical Identifiers

Chemical Name or Material 3,4-Difluoro-5-methylbenzaldehyde
CAS 1017778-64-1
Molecular Formula C8H6F2O
Formula Weight 156.13
MDL Number MFCD09832333
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3198903

  SDS
alfa aesar™
H3198903
1g Each for $275.53

AAH31989MD

  SDS
alfa aesar™
H31989MD
250mg Each for $82.35
Description & Specifications

Specifications

Formula Weight 156.13
Chemical Name or Material 3,4-Difluoro-5-methylbenzaldehyde
Assay 97%
Molecular Formula C8H6F2O
MDL Number MFCD09832333
CAS 1017778-64-1
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.