missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Difluoro-4'-(4-ethylcyclohexyl)biphenyl, 97%

Manufacturer:  Alfa Aesar™ H5527506

 View more versions of this product

Catalog No. AAH5527506



Specifications

Specifications

3,4-Difluoro-4'-(4-ethylcyclohexyl)biphenyl
5g
C20H22F2
1,1'-biphenyl, 4'-4-ethylcyclohexyl-3,4-difluoro
CCC1CCC(CC1)C2=CC=C(C=C2)C3=CC(=C(C=C3)F)F
300.393
300.39
68°C to 69°C
134412-18-3
MFCD12828121
TYZNCUASDJPXMP-UHFFFAOYSA-N
4-[4-(4-ethylcyclohexyl)phenyl]-1,2-difluorobenzene
612308
97%

Chemical Identifiers

134412-18-3
300.393
TYZNCUASDJPXMP-UHFFFAOYSA-N
612308
CCC1CCC(CC1)C2=CC=C(C=C2)C3=CC(=C(C=C3)F)F
C20H22F2
MFCD12828121
1,1'-biphenyl, 4'-4-ethylcyclohexyl-3,4-difluoro
4-[4-(4-ethylcyclohexyl)phenyl]-1,2-difluorobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

Documents