missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Difluoro-4'-(4-ethylcyclohexyl)biphenyl, 97%

Manufacturer:  Alfa Aesar™ H5527503

 View more versions of this product

Catalog No. AAH5527503



Specifications

Specifications

3,4-Difluoro-4'-(4-ethylcyclohexyl)biphenyl
1g
C20H22F2
1,1'-biphenyl, 4'-4-ethylcyclohexyl-3,4-difluoro
CCC1CCC(CC1)C2=CC=C(C=C2)C3=CC(=C(C=C3)F)F
300.393
300.39
68°C to 69°C
134412-18-3
MFCD12828121
TYZNCUASDJPXMP-UHFFFAOYSA-N
4-[4-(4-ethylcyclohexyl)phenyl]-1,2-difluorobenzene
612308
97%

Chemical Identifiers

134412-18-3
300.393
TYZNCUASDJPXMP-UHFFFAOYSA-N
612308
CCC1CCC(CC1)C2=CC=C(C=C2)C3=CC(=C(C=C3)F)F
C20H22F2
MFCD12828121
1,1'-biphenyl, 4'-4-ethylcyclohexyl-3,4-difluoro
4-[4-(4-ethylcyclohexyl)phenyl]-1,2-difluorobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

TSCA : No

Recommended Storage : Ambient temperatures

Documents