Please login to your online account to display your discounted pricing

3,4-Difluoro-2-methylbenzoic acid, 97+%, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material 3,4-Difluoro-2-methylbenzoic acid
CAS 157652-31-8
Molecular Formula C8H6F2O2
Formula Weight 172.13
MDL Number MFCD07777153
Synonym 3,4-Difluoro-o-toluic acid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAH3232603

  SDS
alfa aesar
H3232603
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3232606

  SDS
alfa aesar
H3232606
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 172.13
Quantity 1g
Assay ≥97%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point 152° to 156°C
MDL Number MFCD07777153
CAS 157652-31-8
Molecular Formula C8H6F2O2
Color White to Cream
Chemical Name or Material 3,4-Difluoro-2-methylbenzoic acid
Synonym 3,4-Difluoro-o-toluic acid
Odor Odorless
Physical Form Powder
Health Hazard 1 Warning

Applications
3,4-Difluoro-2-methylbenzoic acid, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used in medicine field, electronic industry and polymer material.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.