missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dichlorotoluene, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1076330

 View more versions of this product

Catalog No. AAA1076330


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3,4-Dichlorotoluene
∼-15°C
200°C to 202°C
C7H6Cl2
MFCD00000556
UN3082
3,4-dichlorotoluene, benzene, 1,2-dichloro-4-methyl, toluene, 3,4-dichloro, 3,4-dichloro-toluene, ksc486q9r, chembl47525, benzene,2-dichloro-4-methyl, 1,2-dichloro-4-methyl-benzene, #
CC1=CC(=C(C=C1)Cl)Cl
161.025
161.03
95-75-0
1.251
85°C (185°F)
1.547
250 g
1931687
WYUIWKFIFOJVKW-UHFFFAOYSA-N
1,2-dichloro-4-methylbenzene
7256
98%

Chemical Identifiers

95-75-0
161.025
WYUIWKFIFOJVKW-UHFFFAOYSA-N
7256
CC1=CC(=C(C=C1)Cl)Cl
C7H6Cl2
MFCD00000556
3,4-dichlorotoluene, benzene, 1,2-dichloro-4-methyl, toluene, 3,4-dichloro, 3,4-dichloro-toluene, ksc486q9r, chembl47525, benzene,2-dichloro-4-methyl, 1,2-dichloro-4-methyl-benzene, #
1,2-dichloro-4-methylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H227-H302-H315-H319-H335

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

EINECSNumber : 202-447-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only