Learn More
3,4-Dichlorophenylhydrazine hydrochloride, 98+%, Thermo Scientific Chemicals
$136.59 - $1639.71
Chemical Identifiers
CAS | 19763-90-7 |
---|---|
Molecular Formula | C6H7Cl3N2 |
Molecular Weight (g/mol) | 213.486 |
MDL Number | MFCD00012937 |
InChI Key | WIKZRYSGQPZTMM-UHFFFAOYSA-N |
Synonym | 3,4-dichlorophenylhydrazine hydrochloride, 3,4-dichlorophenyl hydrazine hydrochloride, 1-3,4-dichlorophenyl hydrazine hydrochloride, 3,4-dichlorophenyl hydrazine monohydrochloride, hydrazine, 3,4-dichlorophenyl-, monohydrochloride, 3,4-dichlorophenylhydrazinehydrochloride, pubchem7530, acmc-209f1i, ksc491e2j |
PubChem CID | 3084323 |
IUPAC Name | (3,4-dichlorophenyl)hydrazine;hydrochloride |
SMILES | C1=CC(=C(C=C1NN)Cl)Cl.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1390609
|
Thermo Scientific Chemicals
A1390609 |
10 g |
Each for $136.59
|
|
|||||
AAA1390618
|
Thermo Scientific Chemicals
A1390618 |
50 g |
Each for $421.04
|
|
|||||
AAA1390630
|
Thermo Scientific Chemicals
A1390630 |
250 g |
Each for $1,639.71
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
19763-90-7 | |
213.486 | |
WIKZRYSGQPZTMM-UHFFFAOYSA-N | |
3084323 | |
C1=CC(=C(C=C1NN)Cl)Cl.Cl |
C6H7Cl3N2 | |
MFCD00012937 | |
3,4-dichlorophenylhydrazine hydrochloride, 3,4-dichlorophenyl hydrazine hydrochloride, 1-3,4-dichlorophenyl hydrazine hydrochloride, 3,4-dichlorophenyl hydrazine monohydrochloride, hydrazine, 3,4-dichlorophenyl-, monohydrochloride, 3,4-dichlorophenylhydrazinehydrochloride, pubchem7530, acmc-209f1i, ksc491e2j | |
(3,4-dichlorophenyl)hydrazine;hydrochloride |
Specifications
19763-90-7 | |
C6H7Cl3N2 | |
10 g | |
3,4-dichlorophenylhydrazine hydrochloride, 3,4-dichlorophenyl hydrazine hydrochloride, 1-3,4-dichlorophenyl hydrazine hydrochloride, 3,4-dichlorophenyl hydrazine monohydrochloride, hydrazine, 3,4-dichlorophenyl-, monohydrochloride, 3,4-dichlorophenylhydrazinehydrochloride, pubchem7530, acmc-209f1i, ksc491e2j | |
C1=CC(=C(C=C1NN)Cl)Cl.Cl | |
213.486 | |
213.5 | |
3,4-Dichlorophenylhydrazine hydrochloride |
∼229°C (decomposition) | |
MFCD00012937 | |
3703934 | |
WIKZRYSGQPZTMM-UHFFFAOYSA-N | |
(3,4-dichlorophenyl)hydrazine;hydrochloride | |
3084323 | |
≥98% |
Safety and Handling
GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
EINECSNumber : 243-278-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only