missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dichlorophenylboronic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 151169-75-4
Molecular Formula C6H5BCl2O2
Molecular Weight (g/mol) 190.81
MDL Number MFCD01074646
InChI Key JKIGHOARKAIPJI-UHFFFAOYSA-N
Synonym 3,4-dichloro benzene boronic acid, 3,4-dichlorobenzeneboronic acid, 3,4-dichlorophenyl boranediol, 3,4-dichlorophenyl boronic acid, 3,4-dichlorophenylbornic acid, 3,4-dichlorophenylboronicacid, boronic acid, 3,4-dichlorophenyl, chembl20776, pubchem1812
PubChem CID 2734330
IUPAC Name (3,4-dichlorophenyl)boronic acid
SMILES OB(O)C1=CC=C(Cl)C(Cl)=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC359090050
SDS View Documents Promotion Details
Thermo Fisher Scientific
AC359090050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

151169-75-4
190.81
JKIGHOARKAIPJI-UHFFFAOYSA-N
2734330
OB(O)C1=CC=C(Cl)C(Cl)=C1
C6H5BCl2O2
MFCD01074646
3,4-dichloro benzene boronic acid, 3,4-dichlorobenzeneboronic acid, 3,4-dichlorophenyl boranediol, 3,4-dichlorophenyl boronic acid, 3,4-dichlorophenylbornic acid, 3,4-dichlorophenylboronicacid, boronic acid, 3,4-dichlorophenyl, chembl20776, pubchem1812
(3,4-dichlorophenyl)boronic acid

Specifications

151169-75-4
100.0
284.0°C to 286.0°C
Authentic
MFCD01074646
3,4-dichloro benzene boronic acid, 3,4-dichlorobenzeneboronic acid, 3,4-dichlorophenyl boranediol, 3,4-dichlorophenyl boronic acid, 3,4-dichlorophenylbornic acid, 3,4-dichlorophenylboronicacid, boronic acid, 3,4-dichlorophenyl, chembl20776, pubchem1812
JKIGHOARKAIPJI-UHFFFAOYSA-N
(3,4-dichlorophenyl)boronic acid
2734330
Powder
3, 4-Dichlorophenylboronic acid
96.0
White
96% min. (HPLC)
C6H5BCl2O2
Glass bottle
5g
OB(O)C1=CC=C(Cl)C(Cl)=C1
190.81
190.82
97%
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre

GHS Signal Word: Warning

SDS
Product Certifications

RUO – Research Use Only