Please login to your online account to display your discounted pricing

3,4-Dichlorophenylacetonitrile, 98%, ACROS Organics™

$75.16 - $226.49

Chemical Identifiers

Chemical Name or Material 3, 4-Dichlorophenylacetonitrile
Name Note 98%
CAS 3218-49-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H5Cl2N
Linear Formula Cl2C6H3CH2CN
Formula Weight 186.04
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC213940250

  SDS
thermo fisher scientific
213940250
Glass bottle 25g Each for $75.16

AC213941000

  SDS
thermo fisher scientific
213941000
Glass bottle 100g Each for $226.49
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 186.04
Quantity 25g
Color White
EINECS Number 221-743-9
TSCA TSCA
Melting Point 38° to 41°C
Beilstein 09, III, 2272
CAS Max % 100.0
Synonym 3, 4-Dichlorobenzyl cyanide
Chemical Name or Material 3, 4-Dichlorophenylacetonitrile
Infrared Spectrum Authentic
Name Note 98%
Assay Percent Range 98%
Molecular Formula C8H5Cl2N
Linear Formula Cl2C6H3CH2CN
MDL Number MFCD00001909
Packaging Glass bottle
CAS 3218-49-3
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.