Please login to your online account to display your discounted pricing

3,4-Dichlorophenylacetone, 99%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  350830010

 View more versions of this product

Catalog No. AC350830010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 203.07
Quantity 1g
Color Clear light yellow
TSCA TSCA
CAS Max % 100.0
Infrared Spectrum Authentic
Refractive Index 1.5510 to 1.5530 (20°C, 589nm)
Chemical Name or Material 3, 4-Dichlorophenylacetone
Name Note 99%
Assay Percent Range 99%
Molecular Formula C9H8Cl2O
MDL Number MFCD00210406
Packaging Glass bottle
CAS 6097-32-1
CAS Min % 98.5

Specifications

Chemical Name or Material 3, 4-Dichlorophenylacetone
Name Note 99%
CAS 6097-32-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H8Cl2O
Formula Weight 203.07
MDL Number MFCD00210406