missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dichlorophenylacetone, 99%, ACROS Organics™

Chemical Identifiers

CAS 6097-32-1
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C9H8Cl2O
Formula Weight 203.07
MDL Number MFCD00210406
Chemical Name or Material 3, 4-Dichlorophenylacetone, 99%
Assay ≥98.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC350830010 SDS Thermo Fisher Scientific
350830010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

6097-32-1
100.0
C9H8Cl2O
MFCD00210406
≥98.5%
98.5
99%
203.07
3, 4-Dichlorophenylacetone, 99%

Specifications

Clear light yellow
6097-32-1
100.0
Liquid
C9H8Cl2O
MFCD00210406
1.5510 to 1.5530 (20°C, 589nm)
≥98.5%
1g
98.5
Authentic
99%
203.07
Glass bottle
3, 4-Dichlorophenylacetone, 99%

Safety and Handling

TSCA : TSCA