missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dichlorophenylacetic acid, 98%, ACROS Organics™

$70.38 - $70.38

Chemical Identifiers

CAS 5807-30-7
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C8H6Cl2O2
Linear Formula ClC6H3CH2CO2H
Formula Weight 205.04
MDL Number MFCD00004333
Chemical Name or Material 3, 4-Dichlorophenylacetic acid, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC279100250 SDS Thermo Fisher Scientific
279100250
Glass bottle 25g Each for $70.38
Specifications Description & Specifications

Chemical Identifiers

5807-30-7
100.0
C8H6Cl2O2
205.04
3, 4-Dichlorophenylacetic acid, 98%
97.5
98%
ClC6H3CH2CO2H
MFCD00004333
≥97.5%

Specifications

White to light yellow
25g
97.5
Authentic
98%
ClC6H3CH2CO2H
MFCD00004333
3, 4-Dichlorophenylacetic acid, 98%
86° to 90°C
5807-30-7
100.0
Crystalline Powder
C8H6Cl2O2
205.04
Glass bottle
≥97.5%

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 227-368-7

TSCA : TSCA