Please login to your online account to display your discounted pricing

3,4-Dichlorophenyl isocyanate, 99+%, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  292230500

 View more versions of this product

Catalog No. AC292230500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Low Melting Solid
Formula Weight 188.01
Quantity 50g
Color White
Melting Point 40° to 43°C
RTECS Number NQ8760000
TSCA TSCA
EINECS Number 203-026-2
Beilstein 12, 1405
CAS Min % 99.0
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Fatal if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 99+%
Linear Formula Cl2C6H3NCO
Molecular Formula C7H3Cl2NO
CAS 102-36-3
Chemical Name or Material 3, 4-Dichlorophenyl isocyanate
Name Note 99+%
MDL Number MFCD00002017

Specifications

Chemical Name or Material 3, 4-Dichlorophenyl isocyanate
Name Note 99+%
CAS 102-36-3
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C7H3Cl2NO
Linear Formula Cl2C6H3NCO
Formula Weight 188.01
MDL Number MFCD00002017
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Fatal if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents