Please login to your online account to display your discounted pricing

3,4-Dichlorophenyl isocyanate, 99+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3, 4-Dichlorophenyl isocyanate
Name Note 99+%
CAS 102-36-3
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C7H3Cl2NO
Linear Formula Cl2C6H3NCO
Formula Weight 188.01
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC292230500

  SDS
thermo fisher scientific
292230500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥99.0%
Physical Form Low Melting Solid
Formula Weight 188.01
Quantity 50g
Color White
RTECS Number NQ8760000
Melting Point 40° to 43°C
EINECS Number 203-026-2
TSCA TSCA
Beilstein 12, 1405
CAS Min % 99.0
CAS Max % 100.0
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Fatal if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 3, 4-Dichlorophenyl isocyanate
Name Note 99+%
Assay Percent Range 99+%
Molecular Formula C7H3Cl2NO
Linear Formula Cl2C6H3NCO
MDL Number MFCD00002017
CAS 102-36-3
Infrared Spectrum Authentic
Packaging Glass bottle
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Fatal if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.