missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dichlorophenoxyacetic acid, 96%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC152980050

 View more versions of this product

Catalog No. AC152980050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3, 4-Dichlorophenoxyacetic acid
White
5g
95.0
96% min. (HPLC)
Cl2C6H3OCH2CO2H
SNYRXHULAWEECU-UHFFFAOYSA-N
2-(3,4-dichlorophenoxy)acetic acid
11495
221.04
96%
Authentic
138.0°C to 142.0°C
588-22-7
100.0
C8H6Cl2O3
2-3,4-dichlorophenoxy acetic acid
C1=CC(=C(C=C1OCC(=O)O)Cl)Cl
221.04
CHEBI:393747
Crystalline Powder

Chemical Identifiers

588-22-7
221.04
2-3,4-dichlorophenoxy acetic acid
CHEBI:393747
C1=CC(=C(C=C1OCC(=O)O)Cl)Cl
C8H6Cl2O3
SNYRXHULAWEECU-UHFFFAOYSA-N
11495
2-(3,4-dichlorophenoxy)acetic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-612-4

SDS Documents