Learn More
3,4-Dichlorophenoxyacetic acid, 96%, Thermo Scientific™
Chemical Identifiers
CAS | 588-22-7 |
---|---|
Molecular Formula | C8H6Cl2O3 |
Molecular Weight (g/mol) | 221.03 |
MDL Number | MFCD00004303 |
InChI Key | SNYRXHULAWEECU-UHFFFAOYSA-N |
Synonym | 3,4-dichlorophenoxyacetic acid, 2-3,4-dichlorophenoxy acetic acid, 3,4-dichlorophenoxy acetic acid, acetic acid, 3,4-dichlorophenoxy, acetic acid,4-dichlorophenoxy, acmc-20anfd, 4-06-00-00954 beilstein handbook reference, 3,4-dichloro-phenoxy-acetic acid, acetic acid,2-3,4-dichlorophenoxy |
PubChem CID | 11495 |
ChEBI | CHEBI:393747 |
IUPAC Name | 2-(3,4-dichlorophenoxy)acetic acid |
SMILES | OC(=O)COC1=CC=C(Cl)C(Cl)=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC152980050
|
Thermo Scientific Chemicals
152980050 |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
588-22-7 | |
221.03 | |
SNYRXHULAWEECU-UHFFFAOYSA-N | |
11495 | |
2-(3,4-dichlorophenoxy)acetic acid |
C8H6Cl2O3 | |
MFCD00004303 | |
3,4-dichlorophenoxyacetic acid, 2-3,4-dichlorophenoxy acetic acid, 3,4-dichlorophenoxy acetic acid, acetic acid, 3,4-dichlorophenoxy, acetic acid,4-dichlorophenoxy, acmc-20anfd, 4-06-00-00954 beilstein handbook reference, 3,4-dichloro-phenoxy-acetic acid, acetic acid,2-3,4-dichlorophenoxy | |
CHEBI:393747 | |
OC(=O)COC1=CC=C(Cl)C(Cl)=C1 |
Specifications
588-22-7 | |
100.0 | |
96% min. (HPLC) | |
Cl2C6H3OCH2CO2H | |
5g | |
SNYRXHULAWEECU-UHFFFAOYSA-N | |
2-(3,4-dichlorophenoxy)acetic acid | |
11495 | |
221.04 | |
3, 4-Dichlorophenoxyacetic acid |
95.0 | |
Authentic | |
C8H6Cl2O3 | |
MFCD00004303 | |
3,4-dichlorophenoxyacetic acid, 2-3,4-dichlorophenoxy acetic acid, 3,4-dichlorophenoxy acetic acid, acetic acid, 3,4-dichlorophenoxy, acetic acid,4-dichlorophenoxy, acmc-20anfd, 4-06-00-00954 beilstein handbook reference, 3,4-dichloro-phenoxy-acetic acid, acetic acid,2-3,4-dichlorophenoxy | |
OC(=O)COC1=CC=C(Cl)C(Cl)=C1 | |
221.03 | |
CHEBI:393747 | |
96% |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 209-612-4
RUO â Research Use Only