missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dichlorophenol, 99%, ACROS Organics™

$53.27 - $53.27

Chemical Identifiers

CAS 95-77-2
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C6H4Cl2O
Linear Formula Cl2C6H3OH
Formula Weight 163
MDL Number MFCD00002258
Chemical Name or Material 3, 4-Dichlorophenol
Assay ≥98.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC113600500 SDS Thermo Fisher Scientific
AC113600500
Glass bottle 50g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC113600100 SDS Thermo Fisher Scientific
113600100
Glass bottle 10g Each for $53.27
Specifications

Description

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Chemical Identifiers

95-77-2
100.0
C6H4Cl2O
163
3, 4-Dichlorophenol
98.5
99%
Cl2C6H3OH
MFCD00002258
≥98.5%

Specifications

White to brownish
163
65° to 68°C
100.0
C6H4Cl2O
99%
98.5
Authentic
≥98.5%
Powder
50g
06, 190
Cl2C6H3OH
95-77-2
MFCD00002258
Glass bottle
3, 4-Dichlorophenol

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning