missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dichlorophenol, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 113600100

 View more versions of this product

Catalog No. AC113600100



Description

Description

Specifications

Specifications

3, 4-Dichlorophenol
Glass bottle
Brown to White
10g
98.5
98.5% min. (GC)
Cl2C6H3OH
06, 190
Solubility in water: slightly soluble
C1=CC(=C(C=C1O)Cl)Cl
163
CHEBI:34323
Powder
Authentic
145°C to 146°C
65°C to 68°C
95-77-2
100.0
C6H4Cl2O
MFCD00002258
070ftm6dvf, 3,3-dichloro-hydroxybenzene, 3,4 dichlorophenol, 3,4-dcp, 3,4-dichlorphenol, 3,4dichlorophenol, 4,5-dichlorophenol, ccris 5904, phenol, 3,4-dichloro, unii-070ftm6dvf
WDNBURPWRNALGP-UHFFFAOYSA-N
3,4-dichlorophenol
7258
163
99%

Chemical Identifiers

95-77-2
163
WDNBURPWRNALGP-UHFFFAOYSA-N
7258
3,4-dichlorophenol
C6H4Cl2O
MFCD00002258
070ftm6dvf, 3,3-dichloro-hydroxybenzene, 3,4 dichlorophenol, 3,4-dcp, 3,4-dichlorphenol, 3,4dichlorophenol, 4,5-dichlorophenol, ccris 5904, phenol, 3,4-dichloro, unii-070ftm6dvf
CHEBI:34323
C1=CC(=C(C=C1O)Cl)Cl
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-450-5

SDS Documents