missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dichloronitrobenzene, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 113530010

 View more versions of this product

Catalog No. AC113530010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

3, 4-Dichloronitrobenzene
Glass bottle
39°C to 45°C
95%
94.0
95%
Cl2C6H3NO2
05, 246
NTBYINQTYWZXLH-UHFFFAOYSA-N
1,2-dichloro-4-nitrobenzene
7443
Crystalline Mass
Authentic
Yellow to Brown
1kg
99-54-7
100.0
C6H3Cl2NO2
MFCD00007207
1,2-dichloro-4-nitro-benzene, 1-nitro-3,4-dichlorobenzene, 3,4-dichlornitrobenzen, 3,4-dichlornitrobenzen czech, 3,4-dichloronitrobenzen czech, 3,4-dichloronitrobenzene, benzene, 1,2-dichloro-4-nitro, ccris 3097, dcnb, unii-ijt74ji7fs
C1=CC(=C(C=C1[N+](=O)[O-])Cl)Cl
191.995
192
≥94%

Chemical Identifiers

99-54-7
191.995
NTBYINQTYWZXLH-UHFFFAOYSA-N
7443
C1=CC(=C(C=C1[N+](=O)[O-])Cl)Cl
C6H3Cl2NO2
MFCD00007207
1,2-dichloro-4-nitro-benzene, 1-nitro-3,4-dichlorobenzene, 3,4-dichlornitrobenzen, 3,4-dichlornitrobenzen czech, 3,4-dichloronitrobenzen czech, 3,4-dichloronitrobenzene, benzene, 1,2-dichloro-4-nitro, ccris 3097, dcnb, unii-ijt74ji7fs
1,2-dichloro-4-nitrobenzene
Safety and Handling

Safety and Handling

GHS P Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Do not eat, drink or smoke when using this product.
Avoid breathing dust/fume/gas/mist/vapors/spray. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 202-764-2

RTECSNumber : CZ5250000

TSCA : TSCA

SDS Documents