missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dichlorocinnamic acid, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC163120050

 View more versions of this product

Catalog No. AC163120050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

3, 4-Dichlorocinnamic acid
White
5g
96.0
96% min. (HPLC)
Cl2C6H3CH=CHCO2H
RRLUFPHCTSFKNR-DUXPYHPUSA-N
(E)-3-(3,4-dichlorophenyl)prop-2-enoic acid
688027
Powder or Crystalline Powder
Authentic
220.0°C to 223.0°C
1202-39-7
100.0
C9H6Cl2O2
2-propenoic acid, 3-3,4-dichlorophenyl, 2e-3-3,4-dichlorophenyl acrylic acid, 2e-3-3,4-dichlorophenyl prop-2-enoic acid, 3',4'-dichlorocinnamic acid, 3,4-dichlorocinnamic acid, 3-3,4-dichlorophenyl acrylic acid, 3-3,4-dichlorophenyl-2-propenoic acid, cinnamic acid, 3,4-dichloro, e-3-3,4-dichlorophenyl acrylic acid, e-3-3,4-dichlorophenyl prop-2-enoic acid
C1=CC(=C(C=C1C=CC(=O)O)Cl)Cl
217.05
217.05
97%

Chemical Identifiers

1202-39-7
217.05
2-propenoic acid, 3-3,4-dichlorophenyl, 2e-3-3,4-dichlorophenyl acrylic acid, 2e-3-3,4-dichlorophenyl prop-2-enoic acid, 3',4'-dichlorocinnamic acid, 3,4-dichlorocinnamic acid, 3-3,4-dichlorophenyl acrylic acid, 3-3,4-dichlorophenyl-2-propenoic acid, cinnamic acid, 3,4-dichloro, e-3-3,4-dichlorophenyl acrylic acid, e-3-3,4-dichlorophenyl prop-2-enoic acid
(E)-3-(3,4-dichlorophenyl)prop-2-enoic acid
C9H6Cl2O2
RRLUFPHCTSFKNR-DUXPYHPUSA-N
688027
C1=CC(=C(C=C1C=CC(=O)O)Cl)Cl
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 214-866-4

SDS Documents