missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dichlorobenzyl isothiocyanate, 97%

Manufacturer:  Alfa Aesar™ L1170103

 View more versions of this product

Catalog No. AAL1170103



Specifications

Specifications

3,4-Dichlorobenzyl isothiocyanate
130°C (1mmHg)
1g
C8H5Cl2NS
UN2923
3,4-dichlorobenzyl isothiocyanate, 1,2-dichloro-4-isothiocyanatomethyl benzene, isothiocyanic acid, 3,4-dichlorobenzyl ester, benzene, 1,2-dichloro-4-isothiocyanatomethyl, acmc-1bqjy, wln: scn1r cg dg, 4-12-00-02393 beilstein handbook reference, 3,4-dichlorobenzylisothiocyanate, 3,4-dichlorophenyl methanisothiocyanate, isothiocyanic acid,4-dichlorobenzyl ester
C1=CC(=C(C=C1CN=C=S)Cl)Cl
218.095
218.11
Irritating
26°C to 28°C
18967-42-5
MFCD00041113
2937608
DPSSHXWTNIGVMQ-UHFFFAOYSA-N
1,2-dichloro-4-(isothiocyanatomethyl)benzene
29350
97%

Chemical Identifiers

18967-42-5
218.095
DPSSHXWTNIGVMQ-UHFFFAOYSA-N
29350
C1=CC(=C(C=C1CN=C=S)Cl)Cl
C8H5Cl2NS
MFCD00041113
3,4-dichlorobenzyl isothiocyanate, 1,2-dichloro-4-isothiocyanatomethyl benzene, isothiocyanic acid, 3,4-dichlorobenzyl ester, benzene, 1,2-dichloro-4-isothiocyanatomethyl, acmc-1bqjy, wln: scn1r cg dg, 4-12-00-02393 beilstein handbook reference, 3,4-dichlorobenzylisothiocyanate, 3,4-dichlorophenyl methanisothiocyanate, isothiocyanic acid,4-dichlorobenzyl ester
1,2-dichloro-4-(isothiocyanatomethyl)benzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

RTECSNumber : NX8485000

Documents