Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dichlorobenzyl isothiocyanate, 97%


Quantity 5g

Manufacturer: alfa aesar™  L1170106

 View more versions of this product

Catalog No. AAL1170106

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 218.11
MDL Number MFCD00041113
CAS 18967-42-5
Assay 97%
Health Hazard 2 GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Chemical Name or Material 3,4-Dichlorobenzyl isothiocyanate
Quantity 5g
Beilstein 2937608
Molecular Formula C8H5Cl2NS
Health Hazard 1 Danger
RTECS Number NX8485000
Odor Irritating
Boiling Point 130°C (1mmHg)
UN Number UN2923
Melting Point 26° to 28°C
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Specifications

Chemical Name or Material 3,4-Dichlorobenzyl isothiocyanate
CAS 18967-42-5
Molecular Formula C8H5Cl2NS
Formula Weight 218.11
MDL Number MFCD00041113
UN Number UN2923
Beilstein 2937608
Accessories

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

SDS & Safety Documents