missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dichlorobenzyl isocyanate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 319220010

 View more versions of this product

Catalog No. AC319220010


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3, 4-Dichlorobenzyl isocyanate
97.5
Colorless
282°C
Authentic
Glass bottle
1.5656 to 1.5676
1 g
3,4-dichlorobenzyl isocyanate, 3,4-dichlorobenzylisocyanate, 1,2-dichloro-4-isocyanatomethyl benzene, benzene, 1,2-dichloro-4-isocyanatomethyl, 1,2-dichloro-4-isocyanatomethyl-benzene, 3,4-dichlorophenyl methanisocyanate, zlchem 123, acmc-20anfi, pubchem2096, 3,4-dichloro-benzyl isocyanate
C1=CC(=C(C=C1CN=C=O)Cl)Cl
202.04
202.04
Slightly Viscous Liquid
19752-09-1
100.0
1.3750g/mL
>110°C
97.5% min. (GC)
C8H5Cl2NO
MFCD00673063
1.375
WLNAHQZFPSSVTP-UHFFFAOYSA-N
1,2-dichloro-4-(isocyanatomethyl)benzene
2733368
98%

Chemical Identifiers

19752-09-1
202.04
WLNAHQZFPSSVTP-UHFFFAOYSA-N
2733368
C1=CC(=C(C=C1CN=C=O)Cl)Cl
C8H5Cl2NO
MFCD00673063
3,4-dichlorobenzyl isocyanate, 3,4-dichlorobenzylisocyanate, 1,2-dichloro-4-isocyanatomethyl benzene, benzene, 1,2-dichloro-4-isocyanatomethyl, 1,2-dichloro-4-isocyanatomethyl-benzene, 3,4-dichlorophenyl methanisocyanate, zlchem 123, acmc-20anfi, pubchem2096, 3,4-dichloro-benzyl isocyanate
1,2-dichloro-4-(isocyanatomethyl)benzene
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only