Learn More
3,4-Dichlorobenzyl chloride, 98+%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 279050250
Specifications
3, 4-Dichlorobenzyl chloride | |
155°C | |
Glass bottle | |
1.41 | |
Colorless to Yellow | |
25g | |
98.0 | |
98% min. (GC) | |
Cl2C6H3CH2Cl | |
05, 300 | |
Solubility in water: .. Other solubilities: soluble in organic solvents | |
C1=CC(=C(C=C1CCl)Cl)Cl | |
195.47 | |
195.47 | |
98+% |
1.4100g/mL | |
Authentic | |
1.5775 | |
122°C to 124°C (14.0mmHg) | |
-3°C | |
102-47-6 | |
100.0 | |
C7H5Cl3 | |
MFCD00000906 | |
1,2-dichloro-4-chloromethyl benzene, 1,2-dichloro-4-chloromethyl-benzene, 3,4-dichlorobenzyl chloride, 3,4-dichlorobenzylchloride, 3,4-dichlorophenylmethyl chloride, 451pb8smie, alpha,3,4-trichlorotoluene, benzene, 1,2-dichloro-4-chloromethyl, toluene, .alpha.,3,4-trichloro, unii-451pb8smie | |
YZIFVWOCPGPNHB-UHFFFAOYSA-N | |
1,2-dichloro-4-(chloromethyl)benzene | |
7609 | |
Liquid |
Chemical Identifiers
102-47-6 | |
195.47 | |
YZIFVWOCPGPNHB-UHFFFAOYSA-N | |
7609 | |
C1=CC(=C(C=C1CCl)Cl)Cl |
C7H5Cl3 | |
MFCD00000906 | |
1,2-dichloro-4-chloromethyl benzene, 1,2-dichloro-4-chloromethyl-benzene, 3,4-dichlorobenzyl chloride, 3,4-dichlorobenzylchloride, 3,4-dichlorophenylmethyl chloride, 451pb8smie, alpha,3,4-trichlorotoluene, benzene, 1,2-dichloro-4-chloromethyl, toluene, .alpha.,3,4-trichloro, unii-451pb8smie | |
1,2-dichloro-4-(chloromethyl)benzene |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 203-033-