Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dichlorobenzyl alcohol, 98%

Chemical Identifiers

Chemical Name or Material 3,4-Dichlorobenzyl alcohol
CAS 1805-32-9
Molecular Formula C7H6Cl2O
Formula Weight 177.03
MDL Number MFCD00004633
UN Number UN2811
Beilstein 2082362
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1323609

  SDS
alfa aesar™
A1323609
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1323630

  SDS
alfa aesar™
A1323630
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1323618

  SDS
alfa aesar™
A1323618
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 3,4-Dichlorobenzyl alcohol
Quantity 10g
CAS 1805-32-9
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H311-H302-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Assay 98%
Molecular Formula C7H6Cl2O
Formula Weight 177.03
MDL Number MFCD00004633
UN Number UN2811
Boiling Point 130°C (3mmHg)
Beilstein 2082362
Melting Point 36° to 39°C
Flash Point >110°C (230°F)
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H311-H302-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.