missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dichlorobenzoylacetonitrile, 96%

$100.08 - $381.46

Chemical Identifiers

CAS 4640-68-0
Molecular Formula C9H5Cl2NO
Formula Weight 214.05
MDL Number MFCD00067919
Beilstein 3267648
Chemical Name or Material 3,4-Dichlorobenzoylacetonitrile
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1072003 SDS Alfa Aesar™
L1072003
1g Each for $100.08
AAL1072006 SDS Alfa Aesar™
L1072006
5g Each for $381.46
Specifications Description & Specifications

Chemical Identifiers

4640-68-0
214.05
3267648
C9H5Cl2NO
MFCD00067919
3,4-Dichlorobenzoylacetonitrile

Specifications

1g
C9H5Cl2NO
MFCD00067919
114° to 117°C
96%
4640-68-0
214.05
3267648
3,4-Dichlorobenzoylacetonitrile

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger