missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dichlorobenzoylacetonitrile, 96%

Manufacturer:  Alfa Aesar™ L1072006

 View more versions of this product

Catalog No. AAL1072006



Specifications

Specifications

3,4-Dichlorobenzoylacetonitrile
5g
C9H5Cl2NO
3267648
HUEULXLHYYYTTP-UHFFFAOYSA-N
3-(3,4-dichlorophenyl)-3-oxopropanenitrile
2758063
96%
114°C to 117°C
4640-68-0
MFCD00067919
3,4-dichlorobenzoylacetonitrile, 3-3,4-dichlorophenyl-3-oxopropanenitrile, 3-3,4-dichloro-phenyl-3-oxo-propionitrile, pubchem12008, acmc-1aspw, dichlorophenyloxopropanenitrile, 3-3,4-dichlorophenyl-3-oxopropionitrile
C1=CC(=C(C=C1C(=O)CC#N)Cl)Cl
214.045
214.05

Chemical Identifiers

4640-68-0
214.045
HUEULXLHYYYTTP-UHFFFAOYSA-N
2758063
C1=CC(=C(C=C1C(=O)CC#N)Cl)Cl
C9H5Cl2NO
MFCD00067919
3,4-dichlorobenzoylacetonitrile, 3-3,4-dichlorophenyl-3-oxopropanenitrile, 3-3,4-dichloro-phenyl-3-oxo-propionitrile, pubchem12008, acmc-1aspw, dichlorophenyloxopropanenitrile, 3-3,4-dichlorophenyl-3-oxopropionitrile
3-(3,4-dichlorophenyl)-3-oxopropanenitrile
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Documents