missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dichlorobenzoyl chloride, 97%, Thermo Scientific Chemicals

$214.67 - $214.67

Chemical Identifiers

CAS 3024-72-4
Molecular Formula C7H3Cl3O
Molecular Weight (g/mol) 209.45
MDL Number MFCD00000672
InChI Key VTXNOVCTHUBABW-UHFFFAOYSA-N
PubChem CID 76403
IUPAC Name 3,4-dichlorobenzoyl chloride
SMILES C1=CC(=C(C=C1C(=O)Cl)Cl)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1700622
View Documents Promotion Details
Thermo Scientific Chemicals
A1700622
100 g
Each for $214.67
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3024-72-4
209.45
VTXNOVCTHUBABW-UHFFFAOYSA-N
3,4-dichlorobenzoyl chloride
C7H3Cl3O
MFCD00000672
76403
C1=CC(=C(C=C1C(=O)Cl)Cl)Cl

Specifications

3024-72-4
242°C
C7H3Cl3O
100 g
607485
benzoyl chloride, 3,4-dichloro, 3,4-dichlorobenzoylchloride, 3,4-dichloro benzoyl chloride, 2,3-dichlorobenzoyl, 3,4dichlorobenzoyl chloride, acmc-1aj45, 3,4 dichlorobenzoyl chloride, 3,4-dichlorobenzoyl-chloride, ksc225o5d, 3,4-dichloro-benzoyl chloride
C1=CC(=C(C=C1C(=O)Cl)Cl)Cl
209.45
209.46
3,4-Dichlorobenzoyl chloride
30°C to 34°C
142°C (287°F)
MFCD00000672
UN3261
Moisture sensitive
VTXNOVCTHUBABW-UHFFFAOYSA-N
3,4-dichlorobenzoyl chloride
76403
97%
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 221-177-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only