Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dichlorobenzotrifluoride, 97%

Chemical Identifiers

Chemical Name or Material 3,4-Dichlorobenzotrifluoride
CAS 328-84-7
Molecular Formula C7H3Cl2F3
Formula Weight 215
MDL Number MFCD00000555
UN Number UN1760
Beilstein 1950151
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1509809

 
alfa aesar™
A1509809
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1509818

 
alfa aesar™
A1509818
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1509830

 
alfa aesar™
A1509830
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1509836

 
alfa aesar™
A1509836
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 215
Chemical Name or Material 3,4-Dichlorobenzotrifluoride
TSCA TSCA
Assay 97%
Molecular Formula C7H3Cl2F3
MDL Number MFCD00000555
CAS 328-84-7
UN Number UN1760
Boiling Point 173° to 174°C
Beilstein 1950151
Melting Point −12°C
Flash Point 65°C (149°F)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
RTECS Number CZ5527510
Quantity 10g
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.