missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dichlorobenzotrifluoride, 97%

Manufacturer:  Alfa Aesar™ A1509836

 View more versions of this product

Catalog No. AAA1509836



Specifications

Specifications

3,4-Dichlorobenzotrifluoride
173°C to 174°C
500g
C7H3Cl2F3
UN1760
3,4-dichlorobenzotrifluoride, 1,2-dichloro-4-trifluoromethyl benzene, benzene, 1,2-dichloro-4-trifluoromethyl, 3,4-dichloro-alpha,alpha,alpha-trifluorotoluene, 3,4-dichlorotrifluoromethylbenzene, unii-7g60xcu4gg, 3,4-dichlorophenyltrifluoromethane, 7g60xcu4gg, 3,4-dichloro-1-trifluoromethyl benzene, 3,4-dcbte
C1=CC(=C(C=C1C(F)(F)F)Cl)Cl
214.996
215
65°C (149°F)
-12°C
328-84-7
MFCD00000555
1950151
XILPLWOGHPSJBK-UHFFFAOYSA-N
1,2-dichloro-4-(trifluoromethyl)benzene
9481
97%

Chemical Identifiers

328-84-7
214.996
XILPLWOGHPSJBK-UHFFFAOYSA-N
9481
C1=CC(=C(C=C1C(F)(F)F)Cl)Cl
C7H3Cl2F3
MFCD00000555
3,4-dichlorobenzotrifluoride, 1,2-dichloro-4-trifluoromethyl benzene, benzene, 1,2-dichloro-4-trifluoromethyl, 3,4-dichloro-alpha,alpha,alpha-trifluorotoluene, 3,4-dichlorotrifluoromethylbenzene, unii-7g60xcu4gg, 3,4-dichlorophenyltrifluoromethane, 7g60xcu4gg, 3,4-dichloro-1-trifluoromethyl benzene, 3,4-dcbte
1,2-dichloro-4-(trifluoromethyl)benzene
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.

RTECSNumber : CZ5527510

TSCA : TSCA

Documents