Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dichlorobenzophenone, 99%

$78.04 - $860.46

Chemical Identifiers

Chemical Name or Material 3,4-Dichlorobenzophenone
CAS 6284-79-3
Molecular Formula C13H8Cl2O
Formula Weight 251.11
MDL Number MFCD00000552
Beilstein 2105069
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2294714

 
alfa aesar™
B2294714
25g Each for $78.04

AAB2294722

 
alfa aesar™
B2294722
100g Each for $204.73

AAB2294736

 
alfa aesar™
B2294736
500g Each for $860.46
Description & Specifications

Specifications

Formula Weight 251.11
Chemical Name or Material 3,4-Dichlorobenzophenone
Odor Odorless
Assay 99%
Molecular Formula C13H8Cl2O
MDL Number MFCD00000552
CAS 6284-79-3
Beilstein 2105069
Melting Point 100° to 104°C
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.