missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dichlorobenzonitrile, 97%

Manufacturer:  Alfa Aesar™ A1233822

 View more versions of this product

Catalog No. AAA1233822


 

Specifications

Specifications

3,4-Dichlorobenzonitrile
6574-99-8
C7H3Cl2N
UN3439
69°C to 74°C
C1=CC(=C(C=C1C#N)Cl)Cl
172.008
172.01
100g
97%
MFCD00016379
2326352
KUWBYWUSERRVQP-UHFFFAOYSA-N
3,4-dichlorobenzonitrile
81051

Chemical Identifiers

6574-99-8
172.008
KUWBYWUSERRVQP-UHFFFAOYSA-N
C7H3Cl2N
MFCD00016379
81051
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

SDS Documents