Please login to your online account to display your discounted pricing

3,4-Dichlorobenzoic acid, 98+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3, 4-Dichlorobenzoic acid
CAS 51-44-5
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C7H4Cl2O2
Linear Formula Cl2C6H3CO2H
Formula Weight 191.01
MDL Number MFCD00002492
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC374215000

  SDS
thermo fisher scientific
AC374215000
Plastic bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.0%
Formula Weight 191.01
Physical Form Powder
Quantity 500g
Color White to beige
Melting Point 205° to 209°C
CAS Min % 98.0
CAS Max % 100.0
Chemical Name or Material 3, 4-Dichlorobenzoic acid
Assay Percent Range 98+%
CAS 51-44-5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
MDL Number MFCD00002492
Infrared Spectrum Authentic
Packaging Plastic bottle
Linear Formula Cl2C6H3CO2H
Molecular Formula C7H4Cl2O2
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.