Please login to your online account to display your discounted pricing

3,4-Dichlorobenzoic acid, 98+%, ACROS Organics™


Quantity 500g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC374215000

 View more versions of this product

Catalog No. AC374215000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Formula Weight 191.01
Physical Form Powder
Quantity 500g
Color White to beige
Melting Point 205° to 209°C
CAS Min % 98.0
CAS Max % 100.0
Chemical Name or Material 3, 4-Dichlorobenzoic acid
Assay Percent Range 98+%
CAS 51-44-5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
MDL Number MFCD00002492
Infrared Spectrum Authentic
Packaging Plastic bottle
Linear Formula Cl2C6H3CO2H
Molecular Formula C7H4Cl2O2
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3, 4-Dichlorobenzoic acid
CAS 51-44-5
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C7H4Cl2O2
Linear Formula Cl2C6H3CO2H
Formula Weight 191.01
MDL Number MFCD00002492
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents