Please login to your online account to display your discounted pricing

3,4-Dichlorobenzenethiol, 97%, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  AC162850010

 View more versions of this product

Catalog No. AC162850010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid or Low Melting Solid
Assay ≥96.0%
Formula Weight 179.06
Quantity 1g
Color Cloudy colorless to yellow or white to yellow
Density 1.4g/mL
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Molecular Formula C6H4Cl2S
MDL Number MFCD00004840
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 3, 4-Dichlorobenzenethiol
Assay Percent Range 97%
CAS 5858-17-3
Linear Formula Cl2C6H3SH
Health Hazard 1 GHS Signal Word: Warning

Specifications

Chemical Name or Material 3, 4-Dichlorobenzenethiol
CAS 5858-17-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H4Cl2S
Linear Formula Cl2C6H3SH
Formula Weight 179.06
MDL Number MFCD00004840
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents