Please login to your online account to display your discounted pricing

3,4-Dichlorobenzenethiol, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3, 4-Dichlorobenzenethiol
CAS 5858-17-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H4Cl2S
Linear Formula Cl2C6H3SH
Formula Weight 179.06
MDL Number MFCD00004840
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC162850010

  SDS
thermo fisher scientific
AC162850010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC162850050

  SDS
thermo fisher scientific
AC162850050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 3, 4-Dichlorobenzenethiol
Color Cloudy colorless to yellow or white to yellow
Quantity 1g
CAS 5858-17-3
CAS Min % 96.0
Health Hazard 1 GHS Signal Word: Warning
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Density 1.4g/mL
Infrared Spectrum Authentic
Physical Form Liquid or Low Melting Solid
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H4Cl2S
Linear Formula Cl2C6H3SH
Formula Weight 179.06
MDL Number MFCD00004840
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.