missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dichlorobenzenesulfonyl chloride, 98%

Manufacturer:  Alfa Aesar™ A1457622

 View more versions of this product

Catalog No. AAA1457622



Specifications

Specifications

3,4-Dichlorobenzenesulfonyl chloride
253°C
100g
C6H3Cl3O2S
UN3265
3,4-dichlorobenzene-1-sulfonyl chloride, 3,4-dichlorophenylsulfonyl chloride, 3,4-dichlorobenzenesulphonyl chloride, benzenesulfonyl chloride, 3,4-dichloro, 3,4-dichlorobenzenesulfonylchloride, 3,4-dichloro-benzenesulfonyl chloride, 3,4-dichlorobenzene-sulfonyl chloride, 3,4-dichloro benzene sulfonyl chloride, 3,4-dichlorophenyl chlorosulfone, pubchem5118
C1=CC(=C(C=C1S(=O)(=O)Cl)Cl)Cl
245.498
245.51
>110°C (230°F)
20°C to 22°C
98-31-7
MFCD00041255
1956417
NYIBPWGZGSXURD-UHFFFAOYSA-N
3,4-dichlorobenzenesulfonyl chloride
7383
98%

Chemical Identifiers

98-31-7
245.498
NYIBPWGZGSXURD-UHFFFAOYSA-N
7383
C1=CC(=C(C=C1S(=O)(=O)Cl)Cl)Cl
C6H3Cl3O2S
MFCD00041255
3,4-dichlorobenzene-1-sulfonyl chloride, 3,4-dichlorophenylsulfonyl chloride, 3,4-dichlorobenzenesulphonyl chloride, benzenesulfonyl chloride, 3,4-dichloro, 3,4-dichlorobenzenesulfonylchloride, 3,4-dichloro-benzenesulfonyl chloride, 3,4-dichlorobenzene-sulfonyl chloride, 3,4-dichloro benzene sulfonyl chloride, 3,4-dichlorophenyl chlorosulfone, pubchem5118
3,4-dichlorobenzenesulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

TSCA : TSCA

Documents